BioCam Cell Structure And Function Charts
Cell Membrane Chart, Laminated, 61 cm, 45.7 cm
Catalog # 470006-416
Supplier: BioCam Communications
Supplier #: WC 51
Jump to:
Product Details & Documents
A mixture of photo-micrographs, detailed artwork and diagrams make these charts excellent for supporting your unit on cells and cell division.
- Highly detailed
Explore cells with these charts.
Specifications
- Width:45.7 cm
- Description:Cell Membrane Chart
- Length:61 cm
- Finish:Laminated
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



