Order Entry
United States
Orders LinkContactUsLinkComponent
Replacement Bulbs

Replacement Bulbs

Replacement Bulb, with Bi-Pin Base, 12 V, 15 W
Catalog # 470005-048
Supplier:   VWR
Supplier #:  SE06.0101
Replacement Bulbs
Catalog # 470005-048
Supplier:  VWR
Replacement Bulbs
Replacement Bulbs

Replacement Bulb, with Bi-Pin Base, 12 V, 15 W

Price
$21.31
Log in for availability
Each
Product Details & Documents

Replacement microscope light bulbs are available for a variety of microscopes. If unsure which bulb type is needed, please contact Ward's Science.

Specifications
  • Description:
    Replacement Bulb, with Bi-Pin Base
  • Electrical:
    12 V, 15 W
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.