Variable Speed Centrifuge
Variable Speed Centrifuge
470003-272
Supplier #: C856
A Valuable Addition to Any Laboratory.
- 30 minute timer with bell tone signal
- Latching lid with Safety Switch
- Easily accessible stainless steel rotor
For a clean, complete separation of cellular components yielding a superior serum or plasma specimen, use this powerful variable speed centrifuge. Suction cup rubber feet for stability also reduce vibration. Centrifuge features all UL approved components.
- Description:Variable Speed Centrifuge
- Cat. No.:470003-272
- Speed:300 - 4000 rpm, variable
- Dimensions:254W x 203H x 302L mm
- Capacities:6 tubes, 2-10 ml



