SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
PBS Blocking Buffer, 100 mL
Supplier: Invitrogen
Supplier #: 37580
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
Catalog # PI37580
Supplier: Invitrogen
Marketsource
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
SuperBlock™ Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
- Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
- Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
- Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilising them for drying and storage for later use.
Documents
Certificates
Search by Lot NumberThermo Scientific Reagents for Manual Western Blot Detection Brochure
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Description:PBS Blocking Buffer
- Size:100 mL
- Application:ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization



