Order Entry
United States
Orders LinkContactUsLinkComponent
pH-Indicator Paper, MilliporeSigma

pH-Indicator Paper, MilliporeSigma

pH Box, pH Indicator Paper and Color Scale, 3 rolls, 4.8 m, 0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-4.5-
5.0-5.5-6.0-6.5-7.0-7.5-8.0-8.5-9.0-9.5-
10.0-10.5-11.0-11.5-12.0-12.5-13.0, 0.5 - 13.0, with Acilit® (pH 0.5 - 5.0), Neutralit® (pH 5.5 - 9.0), Alkalit® (9.5 - 13.0)
Catalog # EM1.09565.0001
Supplier:   MilliporeSigma
Supplier #:  1095650001
pH-Indicator Paper, MilliporeSigma
Catalog # EM1.09565.0001
Supplier:  MilliporeSigma
pH-Indicator Paper, MilliporeSigma

pH Box, pH Indicator Paper and Color Scale, 3 rolls, 4.8 m, 0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-4.5-
5.0-5.5-6.0-6.5-7.0-7.5-8.0-8.5-9.0-9.5-
10.0-10.5-11.0-11.5-12.0-12.5-13.0, 0.5 - 13.0, with Acilit® (pH 0.5 - 5.0), Neutralit® (pH 5.5 - 9.0), Alkalit® (9.5 - 13.0)

Price
$85.69
Log in for availability
Each Case of 144
Product Details & Documents

pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences

MilliporeSigma # 1.09562.0003 replaces and is identical to MilliporeSigma # 9562-2.
MilliporeSigma # 1.09564.0003 replaces and is identical to MilliporeSigma # 9564-2.

pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.

Specifications
  • Description:
    pH Box, pH Indicator Paper and Color Scale
  • Length:
    4.8 m
  • Size:
    3 rolls
  • pH range:
    0.5 - 13.0, with Acilit® (pH 0.5 - 5.0), Neutralit® (pH 5.5 - 9.0), Alkalit® (9.5 - 13.0)
  • pH graduation:
    0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-4.5-
    5.0-5.5-6.0-6.5-7.0-7.5-8.0-8.5-9.0-9.5-
    10.0-10.5-11.0-11.5-12.0-12.5-13.0
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.