pH-Indicator Paper, MilliporeSigma
pH-Indicator Paper, Phenolphthalein Paper Roll with Color Scale, 3 rolls, 4.8 m, —, <8.5 colorless/>8.5 red
Supplier: MilliporeSigma
Supplier #: 1095210003
Jump to:
Product Details & Documents
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences
MilliporeSigma # 1.09562.0003 replaces and is identical to MilliporeSigma # 9562-2.
MilliporeSigma # 1.09564.0003 replaces and is identical to MilliporeSigma # 9564-2.
pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.
Specifications
- Description:pH-Indicator Paper, Phenolphthalein Paper Roll with Color Scale
- Length:4.8 m
- Size:3 rolls
- pH range:<8.5 colorless/>8.5 red
- pH graduation:—