Order Entry
United States
Orders LinkContactUsLinkComponent
 

LiChrolut® Polypropylene Extraction Columns, MilliporeSigma

LiChrolut® RP18, RP-18, 40 - 63 µm, 3 ml, 200 mg
Catalog # EM1.02014.0001
Supplier:   MilliporeSigma
 :  1.02014.0001
LiChrolut® Polypropylene Extraction Columns, MilliporeSigma
Catalog # EM1.02014.0001
Supplier:  MilliporeSigma
LiChrolut® Polypropylene Extraction Columns, MilliporeSigma
LiChrolut® Polypropylene Extraction Columns, MilliporeSigma

LiChrolut® RP18, RP-18, 40 - 63 µm, 3 ml, 200 mg

Price
$415.30
Log in for availability
Each
Product Details & Documents

For nonpolar extraction, choose RP-18, RP-18E (Endcapped), or CN (Cyano) columns.

Typical sample matrix: aqueous buffer solution. Typical sample substances: aromatic ring systems and compounds with alkyl chains. Typical elution solvent: acetonitrile, methanol, and ethyl acetate. Endcapped cartridges have a bonded phase that is reacted with trimethylchlorosilane to cover silanols not previously reacted with the initial bonding reagent. For polar extraction, choose CN (Cyano) or Si (Silica Gel) columns. Typical sample matrix: hexane, oils, and chlorinated hydrocarbons. Typical sample substances: hydroxyl groups, amines, and compounds with hetero-atoms (S, N, O). Typical elution solvent: methanol and 2-propanol.

For nonpolar extraction on a polymer phase, choose EN columns. Typical sample matrix: drinking water, ground water, surface water, and body fluids. Typical sample substances: polar contaminants, pesticides, phenols, explosives, anilines, and pharmaceuticals. Typical elution solvent: ethyl acetate, methanol, acetonitrile, and methanol (1:1).

For anion exchange extraction, choose NH₂ (weak) columns. Typical sample matrix: methanolic/aqueous buffer with low ionic strength, 2 pH units over pK value of the sample substance. Typical sample substances: anions, carboxylic acids, sulfonic acids, and phosphates. Typical elution solvent: aqueous buffer of high ionic strength (0.1 mol/L), 2 pH units over pK value of the sample substance.

For cation exchange extraction choose SCX (strong) columns. Typical sample matrix: methanolic/aqueous buffer with low ionic strength, 2 pH units under pK value of the sample substance. Typical sample substances: cations, amines, and pyrimidines. Typical elution solvent: aqueous buffer of high ionic strength (0.1 mol/L), 2 pH units over pK value of the sample substance.

For mixed mode extraction, choose TSC columns. Typical sample matrix: body fluids. Typical sample substances: cationic and neutral analytes. Typical elution solvent: chloroform-acetone, NH₃-ethyl-acetate, and NH₃-methanol.

 : Sample preparation unit, drying unit and other accessories are sold separately.

Specifications
  • Particle Size:
    40 - 63 µm
  • Capacity:
    3 ml
  • Description:
    LiChrolut® RP18
  • Bed weight:
    200 mg
  • Phase:
    RP-18
Accessories

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Sample Preparation Unit
    Sample Preparation Unit
    48219-224
    Runs up to 12 SPE columns simultaneously
    Log in for availability
    Each - $2,679.56

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Drying Unit
    Drying Unit
    48219-226
    Directs an inert gas through each of the 12 positions to dry the column packing before eluting
    Log in for availability
    Each - $1,875.25

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Disposable fluoroplastic liners
    Disposable fluoroplastic liners
    52428-704
    Log in for availability
    Each - $342.71