Nutrient Broth No. 2, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Catalog # 89406-808
Product available on GSA Advantage®
Supplier: Hardy Diagnostics
Supplier #: C8822
Nutrient Broth No. 2, CRITERION™, Hardy Diagnostics
Catalog # 89406-808
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Nutrient Broth No
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
2, used for the cultivation of fastidious and other microorganisms. Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Specifications
Specifications
- Pack type:Bucket
- Size:2 kg
- Cat. No.:89406-808
- Storage Temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



