D/E Neutralizing Broth, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Supplier: Hardy Diagnostics
: C7372
D/E Neutralizing Broth, CRITERION™, Hardy Diagnostics
Catalog # 89406-312
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
CRITERION™ D/E Neutralizing Broth is an employed for neutralizing of antiseptics and disinfectants and detecting organisms remaining after treatment
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
This broth is especially suited for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity. Dehydratedculture media with engineered container.
: BSE Free
Specifications
Specifications
- Pack type:Bucket
- Size:2 kg
- Cat. No.:89406-312
- Storage Temperature:2-8°C



