D/E Neutralizing Broth, CRITERION™, Hardy Diagnostics
122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
89406-308
Product available on GSA Advantage®
: C7370
D/E Neutralizing Broth, CRITERION™, Hardy Diagnostics
89406-308
Product available on GSA Advantage®
:
CRITERION™ D/E Neutralizing Broth is an employed for neutralizing of antiseptics and disinfectants and detecting organisms remaining after treatment
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
This broth is especially suited for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity. Dehydratedculture media with engineered container.
: BSE Free
- Pack type:Zip-pouch to make 2 L, Mylar®
- Size:122.4 g
- Final volume:2 L
- Cat. No.:89406-308
- Storage Temperature:2-8°C



