Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: Hardy Diagnostics
Supplier #: C5571
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Catalog # 89405-364
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Cat. No.:89405-364
- Storage Temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



