Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # 89405-360
Product available on GSA Advantage®
Supplier: Hardy Diagnostics
Supplier #: C5563
Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
Catalog # 89405-360
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Deoxycholate Lactose Agar for cultivation, maintenance, fermentation reaction and motility determination of bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Bucket
- Size:10 kg
- Cat. No.:89405-360
- Storage Temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



