Orders LinkContactUsLinkComponent
U-tek® Ice Packets, Electron Microscopy Sciences
U-tek® Ice Packets, Electron Microscopy Sciences
Model 405, Mat U-tek® Ice Packets
Catalog # 102680-986
Supplier #:  61533-05
U-tek® Ice Packets, Electron Microscopy Sciences
Catalog # 102680-986
U-tek® Ice Packets, Electron Microscopy Sciences
U-tek® Ice Packets, Electron Microscopy Sciences
Model 405, Mat U-tek® Ice Packets
Price
$29.87
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
U-tek® Ice Packets are cushion fragile shipments which won’t dilute ice baths or damage samples
The packets may be refrozen and reuse, withstands pressure of 60 lb per square inch for 5 minutes or longer. The refrigerant contains neutral pH distilled deionized water and proprietary materials certified by the US Food and Drug Administration as food grade chemicals.
-1°C / +30°F
They remain frozen longer than regular ice without the melting mess.
Specifications
  • Description:
    Model 405, Mat U-tek® Ice Packets