Order Entry
United States
Orders LinkContactUsLinkComponent
MEGA FlexTubes, IBI Scientific

MEGA FlexTubes, IBI Scientific

MEGA FlexTube Kit, 15 ml, 12 K
Catalog # 89369-660
Supplier:   IBI Scientific
Supplier #:  IB48441
MEGA FlexTubes, IBI Scientific
Catalog # 89369-660
Supplier:  IBI Scientific
MEGA FlexTubes, IBI Scientific
MEGA FlexTubes, IBI Scientific

MEGA FlexTube Kit, 15 ml, 12 K

Price
$257.34
Log in for availability
Pack of 15
Product Details & Documents

IBI MEGA FlexTubes are typically used for dialysis or buffer exchange with volumes of 3 to 20 ml.

The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.

IBI FlexTubes allow rapid, secure, and simple loading and recovery of samples. IBI FlexTubes can also be used for applications such as the preparation of protein samples for MALDI-MS, sample concentration, tissue culture extraction purification, removal of salts, surfactants, solvents, and detergents, as well as peptide dialysis and virus-particle purification. The performance of IBI MEGA FlexTubes are regularly monitored and tested by simple dialysis and assays commonly used for proteins and/or nucleic acids.

Specifications
  • Volume:
    15 ml
  • Description:
    MEGA FlexTube Kit
  • MWCO:
    12 K
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.