MEGA FlexTubes, IBI Scientific
MEGA FlexTube Kit, Float, 10 ml, 3.5 K
Catalog # 89369-612
Supplier: IBI Scientific
Supplier #: IB48340
Jump to:
Product Details & Documents
IBI MEGA FlexTubes are typically used for dialysis or buffer exchange with volumes of 3 to 20 ml.
The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.
IBI FlexTubes allow rapid, secure, and simple loading and recovery of samples. IBI FlexTubes can also be used for applications such as the preparation of protein samples for MALDI-MS, sample concentration, tissue culture extraction purification, removal of salts, surfactants, solvents, and detergents, as well as peptide dialysis and virus-particle purification. The performance of IBI MEGA FlexTubes are regularly monitored and tested by simple dialysis and assays commonly used for proteins and/or nucleic acids.
Specifications
- Volume:10 ml
- Description:MEGA FlexTube Kit, Float
- MWCO:3.5 K



