Order Entry
United States
Orders LinkContactUsLinkComponent
MIDI FlexTubes, IBI Scientific

MIDI FlexTubes, IBI Scientific

MIDI FlexTube Kit, Float, 800 µl, 50 - 800 µl, 6 - 8 K
Catalog # 89369-468
Supplier:   IBI Scientific
Supplier #:  IB48024
MIDI FlexTubes, IBI Scientific
Catalog # 89369-468
Supplier:  IBI Scientific
MIDI FlexTubes, IBI Scientific
MIDI FlexTubes, IBI Scientific

MIDI FlexTube Kit, Float, 800 µl, 50 - 800 µl, 6 - 8 K

Price
$43.29
Log in for availability
Pack of 2
Product Details & Documents

IBI MIDI FlexTubes are typically used for the extraction of proteins, RNA, DNA, or oligonucleotides (>20 nt) from polyacrylamide, agarose, or any other gel matrix in a running buffer.

MIDI FlexTubes require a minimum of 0.5 µg of protein at the start of extraction. The maximum size of the gel slice that can be inserted into the MIDI FlexTube is 1×0.5 cm.

The yield of DNA/RNA recovery from an agarose gel or polyacrylamide gel is 90%. The yield of protein recovery from an SDS-PAGE gel is 70%. The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.

They are also used for dialysis or buffer exchange of small volumes (50 to 800 µl), preparation of protein samples for MALDI-MS, 2D gel dots extraction, and sample concentrations. IBI MIDI FlexTubes are tested by using them in the extraction of proteins, RNA, or DNA fragments of various sizes from either agarose or polyacrylamide gels. IBI MIDI FlexTube kits are also tested for simple dialysis of salts or buffer exchanges. The quality of the isolated protein, RNA, or DNA fragments, or of the sample after dialysis, is checked by several assays commonly used for proteins, nucleic acids, and dialysis. These devices combine two modes of action, electro-elution of macromolecules from polyacrylamide or agarose gels and buffer exchange of small volume samples. These devices allow rapid run times and high performance in either mode, and extract the macromolecules without contamination.

Specifications
  • Volume:
    800 µl
  • Description:
    MIDI FlexTube Kit, Float
  • MWCO:
    6 - 8 K
  • For sample range:
    50 - 800 µl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.