Order Entry
United States
Orders LinkContactUsLinkComponent
pH Indicator solution, wide range pH 4-10

pH Indicator solution, wide range pH 4-10

Plastic bottle, 1 L
Catalog # 89231-042
Flammable
Irritant
pH Indicator solution, wide range pH 4-10
Catalog # 89231-042
Supplier:  
CAS Number:  
pH Indicator solution, wide range pH 4-10
pH Indicator solution, wide range pH 4-10

Plastic bottle, 1 L

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WIDE RANGE PH IND SOLN 1L Catalog #104086-122

Product 104086-122 is a replacement for 89231-042 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

For visual determination of pH, using test kits

Method: 10222

  • UN:1219
  • ADR:3,II

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L

Specifications