ProtoScript II Reverse Transcriptase, New England Biolabs
ProtoScript II Reverse Transcriptase, 10,000 units
Catalog # 102500-078
Supplier: New England Biolabs (NEB)
Supplier #: M0368L
ProtoScript II Reverse Transcriptase, New England Biolabs
Catalog # 102500-078
Supplier: New England Biolabs (NEB)
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
ProtoScript II Reverse Transcriptase is a recombinant M-MuLV reverse transcriptase with reduced RNase H activity and increased thermostability
It can be used to synthesize first strand cDNA at higher temperatures than the wild type M-MuLV. The enzyme is active up to 50°C, providing higher specificity, higher yield of cDNA and more full length cDNA product, up to 12 kb in length. This product was formerly known as M-MuLV Reverse Transcriptase RNase H–.
Specifications
Specifications
- Description:ProtoScript II Reverse Transcriptase
- Size:10,000 units