Orders LinkContactUsLinkComponent
MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
Single Standard Ho
Catalog # 102099-072
Supplier #:  80093-HO
MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
Catalog # 102099-072
CAS Number:  
MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
Single Standard Ho
CAS Number:  
Price
$551.45
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
MAC X-ray Microanalysis standards are supplied as 33 m (1/4") diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training.
Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials, some of which are harmful
Universal Set of 20 Elements :Na, Mg, Al, Si, P, S, Cl, Ca, Ti, Cr, Mn, Fe, Ni, Cu, Zn, Nb, Sn, Ba, W, Pb
Rare Earth Set of 10 Elements: La, Ce, Pr, Nd, Sm, Eu, Gd, Dy, Ho, Yb
Precious Metal Set of 5 Elements: Rh, Pd, Ag, Pt, Au
The precious metal foils are approximately 0.125 mm thick and are stretched across plastic supports. Standards are available for 60 elements.
Specifications
Specifications
  • Description:
    Single Standard Ho