MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
Product Details & Documents
MAC X-ray Microanalysis standards are supplied as 33 m (1/4") diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training.
Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials, some of which are harmful
Universal Set of 20 Elements :Na, Mg, Al, Si, P, S, Cl, Ca, Ti, Cr, Mn, Fe, Ni, Cu, Zn, Nb, Sn, Ba, W, Pb
Rare Earth Set of 10 Elements: La, Ce, Pr, Nd, Sm, Eu, Gd, Dy, Ho, Yb
Precious Metal Set of 5 Elements: Rh, Pd, Ag, Pt, Au
The precious metal foils are approximately 0.125 mm thick and are stretched across plastic supports. Standards are available for 60 elements.
Specifications
Specifications
- Description:Single Standard Fe