Orders LinkContactUsLinkComponent
Specialty Labels, Electron Microscopy Sciences
Specialty Labels, Electron Microscopy Sciences
Catalog # 102098-692
Specialty Labels, Electron Microscopy Sciences
Catalog # 102098-692
Supplier:  
Specialty Labels, Electron Microscopy Sciences
Specialty Labels, Electron Microscopy Sciences
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Preprinted Tough-Spots make labeling and identifying samples quick and easy. 
  • Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.