Orders LinkContactUsLinkComponent
Large Set Screw Vises, Electron Microscopy Sciences
Large Set Screw Vises, Electron Microscopy Sciences
Large set screw vise, M4; dia. 25× H 10 mm
Catalog # 102098-264
Supplier #:  75941-03H
Large Set Screw Vises, Electron Microscopy Sciences
Catalog # 102098-264
Large Set Screw Vises, Electron Microscopy Sciences
Large Set Screw Vises, Electron Microscopy Sciences
Large set screw vise, M4; dia. 25× H 10 mm
Price
$77.51
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This set screw vise is made from machined aluminum with stainless steel allen set screws has an open slot of diamension 10 W×5 D mm (0.394×0.197").
Specifications
  • Description:
    Large set screw vise, M4; dia. 25× H 10 mm