Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 100, Black
Catalog # 102097-576
Supplier: Electron Microscopy Sciences
Supplier #: 71478-18
Slide Box-Foam, Electron Microscopy Sciences
Catalog # 102097-576
Supplier: Electron Microscopy Sciences
Jump to:
Product Details & Documents
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
- Holds 25, 50, or 100 slides
- Foam backing
- Secure lock
- Blue, green, red, yellow, white, black, and pink
Specifications
- Capacity:100
- Color:Black
- Description:Slide Box