Slide Box-Foam, Electron Microscopy Sciences
Product Details & Documents
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
- Holds 25, 50, or 100 slides
- Foam backing
- Secure lock
- Blue, green, red, yellow, white, black, and pink
Specifications
- Capacity:100
- Color:Red
- Description:Slide Box