Grid Staining Matrix System, Electron Microscopy Sciences
Product Details & Documents
The Matrix system has a simply alpha-numeric identification system. The unit is not solvent or chemically resistant to acids so all stains should be aqueous based only.
- Stains up to 25 grids at one time or as little as one
The system requires very little stain and you may use different vessels for each stain.
: Each system includes the Matrix Body and 2 each of the Staining Vessels (red and blue).
Specifications
- Description:Staining Vessel, Blue