Orders LinkContactUsLinkComponent
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System Kit
Catalog # 102097-452
Supplier #:  71179-01
Grid Staining Matrix System, Electron Microscopy Sciences
Catalog # 102097-452
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System, Electron Microscopy Sciences
Grid Staining Matrix System Kit
Price
$220.64
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Matrix system has a simply alpha-numeric identification system. The unit is not solvent or chemically resistant to acids so all stains should be aqueous based only.
  • Stains up to 25 grids at one time or as little as one
The system requires very little stain and you may use different vessels for each stain.
Delivery information: Each system includes the Matrix Body and 2 each of the Staining Vessels (red and blue).
Specifications
  • Description:
    Grid Staining Matrix System Kit