Orders LinkContactUsLinkComponent
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Capsules, Size 3
Catalog # 102096-534
Supplier #:  69910-05
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Catalog # 102096-534
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Capsules, Size 3
Price
$61.25
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
Specifications
  • Description:
    Embedding Capsules, Size 3
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Opaque Capsule Holder Size 3 Capsule
    Opaque Capsule Holder Size 3 Capsule
    102681-180
    Log in for availability
    Each - $31.20
Product Information
  • Catalog #
    Availability
    Price
  • Opaque Capsule Holder Size 3 Capsule
    Opaque Capsule Holder Size 3 Capsule
    102681-182
    Log in for availability
    Pack of 6 - $135.29
Product Information
  • Catalog #
    Availability
    Price
  • Opaque Capsule Holder Size 3 Capsule
    Opaque Capsule Holder Size 3 Capsule
    102681-184
    Log in for availability
    Pack of 12 - $250.39
Product Information
  • Catalog #
    Availability
    Price
  • BEEM Capsule Holder White for Size 3
    BEEM Capsule Holder White for Size 3
    103301-920
    Log in for availability
    Pack of 1 - $19.82
Product Information
  • Catalog #
    Availability
    Price
  • BEEM Capsule Holder White for Size 3
    BEEM Capsule Holder White for Size 3
    103302-262
    Log in for availability
    Pack of 6 - $114.02
Product Information
  • Catalog #
    Availability
    Price
  • BEEM Capsule Holder White for Size 3
    BEEM Capsule Holder White for Size 3
    103305-110
    Log in for availability
    Each - $213.69