Paraffin Tissue Microarrays, Electron Microscopy Sciences
Product Details & Documents
Paraffin tissue microarrays (TMAs) are becoming more common in the laboratories. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process. Arraymold features include:
- Allow pathologist or researcher to view many different samples on the same single slide
- Helpful in finding IHC markers for control or quality assurance from hundreds of samples
- Staining markers will be more consistent when hundreds of samples are on one slide
- Used less costly antibody
- Small enough to be stored in a drawer
- Rubber arraymold can be reused many times without losing its flexibility
Current techniques including organizing, punching, and inserting tissues is time consuming and involving costly equipments.This Arraymold is the simple tool but great answer for this current technique for laboratories to perform TMAs with less time consuming, excellent results. With this Arraymold, you can process up to 150 specimens onto one single slide with very little time consuming, minimal technical training.
Specifications
- Description:Arraymold package Kit 1
- Includes:Arraymold Kits A & B
Accessories
Product Information
- Catalog #AvailabilityPrice
- Styler 4.0 mm
Product Information
- Catalog #AvailabilityPrice
- Styler 3.5 mm
Product Information
- Catalog #AvailabilityPrice
- Styler 3.0 mm
Product Information
- Catalog #AvailabilityPrice
- Styler 1.5 mm