Orders LinkContactUsLinkComponent
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences
50 large bags for Lab-sorb
Catalog # 102096-346
Supplier #:  68552-13
Lab-Sorb, Electron Microscopy Sciences
Catalog # 102096-346
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences
50 large bags for Lab-sorb
Price
$52.86
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Converts Liquid Waste to Solid Waste for Easy Disposal
Convenient - Includes extra durable disposal bags, super absorbant LabSorb granules and a convenient graduated scoop.
Prevents Spills - Reduces the possibility of accidentally spilling liquid waste in the lab. If spills do occur, use LabSorb for safe liquid containment during cleanup.
Quick - Convert liquids to solids in seconds.
Adaptable - Apply to any volume of liquid waste including 500 mL, 600 mL, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.
Versatile - LabSorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
Specifications
  • Description:
    50 large bags for Lab-sorb