Lab-Sorb, Electron Microscopy Sciences
Product Details & Documents
Converts Liquid Waste to Solid Waste for Easy Disposal
Convenient - Includes extra durable disposal bags, super absorbant LabSorb granules and a convenient graduated scoop.
Prevents Spills - Reduces the possibility of accidentally spilling liquid waste in the lab. If spills do occur, use LabSorb for safe liquid containment during cleanup.
Quick - Convert liquids to solids in seconds.
Adaptable - Apply to any volume of liquid waste including 500 mL, 600 mL, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.
Versatile - LabSorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
Specifications
- Description:Kit Lab-Sorb 16 oz jar plus 25 medium bags