Order Entry
United States
Orders LinkContactUsLinkComponent
Lab-Sorb, Electron Microscopy Sciences

Lab-Sorb, Electron Microscopy Sciences

Lab-Sorb 16 oz jar, 16 oz.
Catalog # 102096-332
Supplier #:  68552-06
Lab-Sorb, Electron Microscopy Sciences
Catalog # 102096-332
Lab-Sorb, Electron Microscopy Sciences
Lab-Sorb, Electron Microscopy Sciences

Lab-Sorb 16 oz jar, 16 oz.

Price
$73.30
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Converts Liquid Waste to Solid Waste for Easy Disposal
Convenient - Includes extra durable disposal bags, super absorbant LabSorb granules and a convenient graduated scoop.
Prevents Spills - Reduces the possibility of accidentally spilling liquid waste in the lab. If spills do occur, use LabSorb for safe liquid containment during cleanup.
Quick - Convert liquids to solids in seconds.
Adaptable - Apply to any volume of liquid waste including 500 mL, 600 mL, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.
Versatile - LabSorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste

Specifications
  • Capacity:
    16 oz.
  • Description:
    Lab-Sorb 16 oz jar

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.