Orders LinkContactUsLinkComponent
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames 24 x 24 cm Cellophane
Catalog # 102096-316
Supplier #:  68511-24
Gel Drying Frames, Electron Microscopy Sciences
Catalog # 102096-316
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames 24 x 24 cm Cellophane
Price
$35.24
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers.
  • Two open frames
  • Two solid back plates
  • 12 side clips
  • 50 sheets of pre-cut cellophane
The 24 x 24 cm frames allow for drying of larger format gels, while the 14 x 14 cm frames are ideal for mini gels and other smaller formats.
Specifications
  • Description:
    Gel Drying Frames 24 x 24 cm Cellophane