Gel Drying Frames, Electron Microscopy Sciences
Product Details & Documents
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers.
- Two open frames
- Two solid back plates
- 12 side clips
- 50 sheets of pre-cut cellophane
The 24 x 24 cm frames allow for drying of larger format gels, while the 14 x 14 cm frames are ideal for mini gels and other smaller formats.
Specifications
- Description:Gel Drying Frames 14 x 14 cm Cellophane