Orders LinkContactUsLinkComponent
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames 14 x 14 cm Gel Frame
  102096-308
 :  68510-14
Gel Drying Frames, Electron Microscopy Sciences
  102096-308
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames 14 x 14 cm Gel Frame
$220.02
Kit of 2
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers.
  • Two open frames
  • Two solid back plates
  • 12 side clips
  • 50 sheets of pre-cut cellophane
The 24 x 24 cm frames allow for drying of larger format gels, while the 14 x 14 cm frames are ideal for mini gels and other smaller formats.
  • Description:
    Gel Drying Frames 14 x 14 cm Gel Frame