USAF Test Charts, Electron Microscopy Sciences
Product Details & Documents
USAF resolution charts are recognized as a universal standard for testing the vertical and horizontal resolution of imaging systems.
Each element on the chart comprises three vertical bars and three horizontal bars, and the detail on these slides is as fine as 0.78 µm (644 line-pairs per mm). The resolution of the imaging system is normally specified as the Group and Element of the finest bars that can be clearly defined.
Specifications
Specifications
- Description:PS75N USAF Test Chart, positive image, groups 2/1 to 9/3 on