Orders LinkContactUsLinkComponent
USAF Test Charts, Electron Microscopy Sciences
USAF Test Charts, Electron Microscopy Sciences
USAF Test Chart, positive image, groups 0/1 to 9/3R75P
Catalog # 102096-162
Supplier #:  68098-03
USAF Test Charts, Electron Microscopy Sciences
Catalog # 102096-162
CAS Number:  
USAF Test Charts, Electron Microscopy Sciences
USAF Test Charts, Electron Microscopy Sciences
USAF Test Chart, positive image, groups 0/1 to 9/3R75P
CAS Number:  
Price
$2,489.08
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
USAF resolution charts are recognized as a universal standard for testing the vertical and horizontal resolution of imaging systems.
Each element on the chart comprises three vertical bars and three horizontal bars, and the detail on these slides is as fine as 0.78 µm (644 line-pairs per mm). The resolution of the imaging system is normally specified as the Group and Element of the finest bars that can be clearly defined.
Specifications
Specifications
  • Description:
    USAF Test Chart, positive image, groups 0/1 to 9/3R75P