Orders LinkContactUsLinkComponent
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
PS4R with NPL Certificate
Catalog # 102096-144
Supplier #:  68047-PS4R-NP
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
Catalog # 102096-144
CAS Number:  
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
PS4R with NPL Certificate
CAS Number:  
Price
$3,160.87
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions.
Specifications
Specifications
  • Description:
    PS4R with NPL Certificate