PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
PS4R with Grats Certificate
Catalog # 102096-140
Supplier: Electron Microscopy Sciences
Supplier #: 68047-PS4R-GR
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
Catalog # 102096-140
Supplier: Electron Microscopy Sciences
CAS Number:
Jump to:
Product Details & Documents
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions.
Specifications
Specifications
- Description:PS4R with Grats Certificate