Tissue Grinder, 0.5 ml, Electron Microscopy Sciences
Product Details & Documents
This unit features a screw threaded neck for use with Wheaton connection®.
- With Teflon® pestle and screw cap
- Made from Wheaton 33 low extractable borosilicate glass
- Autoclavable
The 13-425 cap has an open top and Teflon® faced silicone rubber liner. The clearance between pestle and mortar is 0.002 – 0.006” (0.05 – 0.15 mm).
Specifications
- Description:Extra Pestle