Order Entry
United States
Orders LinkContactUsLinkComponent
Tissue Grinder, 0.5 ml, Electron Microscopy Sciences

Tissue Grinder, 0.5 ml, Electron Microscopy Sciences

Tissue Grinder, GPI 13-425 cap 0.5 ml
Catalog # 102095-702
Supplier #:  64802-05
Tissue Grinder, 0.5 ml, Electron Microscopy Sciences
Catalog # 102095-702
Tissue Grinder, 0.5 ml, Electron Microscopy Sciences
Tissue Grinder, 0.5 ml, Electron Microscopy Sciences

Tissue Grinder, GPI 13-425 cap 0.5 ml

Price
$492.82
Log in for availability
Pack of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This unit features a screw threaded neck for use with Wheaton connection®.

  • With Teflon® pestle and screw cap
  • Made from Wheaton 33 low extractable borosilicate glass
  • Autoclavable

The 13-425 cap has an open top and Teflon® faced silicone rubber liner. The clearance between pestle and mortar is 0.002 – 0.006” (0.05 – 0.15 mm).

Specifications
  • Description:
    Tissue Grinder, GPI 13-425 cap 0.5 ml

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.