Orders LinkContactUsLinkComponent
FlexTainer Containers, Electron Microscopy Sciences
FlexTainer Containers, Electron Microscopy Sciences
8 L, Flex-Tainer™ Accordion-style Jug
Catalog # 102094-964
Supplier #:  64228-18
FlexTainer Containers, Electron Microscopy Sciences
Catalog # 102094-964
FlexTainer Containers, Electron Microscopy Sciences
FlexTainer Containers, Electron Microscopy Sciences
8 L, Flex-Tainer™ Accordion-style Jug
Price
$242.46
Log in for availability
Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This is a space-saving expandable accordion-style jug with flattens down for easy transport and storage.
Ideal for storing and carrying numerous types of fluids, it is suitable for any laboratory. Made of high strength polyethylene, it features comfortable carry grip and screw-on cap with on/off spout. It is available in many sizes up to 8 liters. Come with individually wrapped.
Specifications
  • Capacity:
    8 L
  • Description:
    Flex-Tainer™ Accordion-style Jug