RDO Rapid Decalcifier, Electron Microscopy Sciences
Product Details & Documents
RDO and RDO Gold were developed with the basic premise of making the work routine quicker and easier without sacraficing the quality and integrity of samples. These are remarkably effective multipurpose decacifiers that can be tailored to suit specific lab routine.
- Safe, non toxic and non-corrosive
- Dilutable and translucent
- Microwavable and biodegradable
- Great cellular differentiation
Both RDO and RDO gold provides crisp nuclear staining and definition, give distinguishable chromatin, show great surface decalcification, safe enough to be disposed down to lab sink to drain.
RDO Gold is designated for biopsies which contain minimal amounts of calcium. It requires no filtration and gives stellar morphological representation of bone biopsies. There is no folding of tissues with RDO Gold and it is clear and non-precipitate.
Documents
Specifications
Specifications
- Description:RDO-Gold Decalcifier
- Size:4×1 gal.