Orders LinkContactUsLinkComponent
RDO Rapid Decalcifier, Electron Microscopy Sciences
RDO Rapid Decalcifier, Electron Microscopy Sciences
RDO-Gold Decalcifier, 1 gal.
Catalog # 102094-950
Supplier #:  64144-40
RDO Rapid Decalcifier, Electron Microscopy Sciences
Catalog # 102094-950
CAS Number:  
RDO Rapid Decalcifier, Electron Microscopy Sciences
RDO Rapid Decalcifier, Electron Microscopy Sciences
RDO-Gold Decalcifier, 1 gal.
CAS Number:  
Price
$176.80
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
RDO and RDO Gold were developed with the basic premise of making the work routine quicker and easier without sacraficing the quality and integrity of samples. These are remarkably effective multipurpose decacifiers that can be tailored to suit specific lab routine.
  • Safe, non toxic and non-corrosive
  • Dilutable and translucent
  • Microwavable and biodegradable
  • Great cellular differentiation
Both RDO and RDO gold provides crisp nuclear staining and definition, give distinguishable chromatin, show great surface decalcification, safe enough to be disposed down to lab sink to drain.
RDO Gold is designated for biopsies which contain minimal amounts of calcium. It requires no filtration and gives stellar morphological representation of bone biopsies. There is no folding of tissues with RDO Gold and it is clear and non-precipitate.
Specifications
Specifications
  • Description:
    RDO-Gold Decalcifier
  • Size:
    1 gal.