NUNC™ MaxiSorp™ 96 Well Plates, Electron Microscopy Sciences
Product Details & Documents
NUNC MaxiSorp plates have been the certified* ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing.
- Hydrophilic MaxiSorp™ surface binds antibodies, protein and biomolecules
- Flat bottom, solid polystyrene plates
- Excellent for chemiluminescent, fluorescent and luminescent arrays
- Standard plate footprint is compatible with exiting readers
* Certified for absorption of IgG; CV of less than 5% between wells. All results are ±10% from the mean and background of all well ±0.005 absorbance units from the mean.
Specifications
- Color:White
- Description:Maxisorp™ 96 Well Plates, PS
- Application:Protein & antibody multiplexing, luminescence
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



