Order Entry
United States
Orders LinkContactUsLinkComponent
 

Probe with Eye, Electron Microscopy Sciences

Probe With Eye, 7"
  102094-614
 :  63324-33
Probe with Eye, Electron Microscopy Sciences
  102094-614
Probe with Eye, Electron Microscopy Sciences
Probe with Eye, Electron Microscopy Sciences

Probe With Eye, 7"

 
$3.28
   
Each
 
   
 

Stainless steel, 2 mm diameter. Flat one end with eye.

 
  • Description:
    Probe With Eye
  • Length:
    7"

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.