Orders LinkContactUsLinkComponent
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe™ Kit for flat bottom microplates
  102093-858
 :  61954-40
CoolSafe Deluxe Kits, Electron Microscopy Sciences
  102093-858
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe™ Kit for flat bottom microplates
$517.46
Kit of 2
Each complete CoolSafe Deluxe system includes 1 CoolSafe Deluxe box, 1 Cool Chamber, and 2 Cool Bricks (1°C & –10°C).
  • Additional sample protection
  • Smaller bench-top footprint
  • Improved portability
  • Clear lid for sample visibility
CoolSafe for 0.2 mL PCR plates or tubes and the CoolSafe™ Deluxe box also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
  • Description:
    CoolSafe™ Kit for flat bottom microplates