Orders LinkContactUsLinkComponent
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe™ Kit for 0.5 ml tubes
Catalog # 102093-854
Supplier #:  61954-20
CoolSafe Deluxe Kits, Electron Microscopy Sciences
Catalog # 102093-854
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe™ Kit for 0.5 ml tubes
Price
$569.20
Log in for availability
Kit of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each complete CoolSafe Deluxe system includes 1 CoolSafe Deluxe box, 1 Cool Chamber, and 2 Cool Bricks (1°C & –10°C).
  • Additional sample protection
  • Smaller bench-top footprint
  • Improved portability
  • Clear lid for sample visibility
CoolSafe for 0.2 mL PCR plates or tubes and the CoolSafe™ Deluxe box also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
Specifications
  • Description:
    CoolSafe™ Kit for 0.5 ml tubes