Orders LinkContactUsLinkComponent
Cell Stretcher, Electron Microscopy Sciences
Cell Stretcher, Electron Microscopy Sciences
Catalog # 102090-744
Cell Stretcher, Electron Microscopy Sciences
Catalog # 102090-744
Supplier:  
Cell Stretcher, Electron Microscopy Sciences
Cell Stretcher, Electron Microscopy Sciences
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cell Stretcher CS-10 Series is a device for simultaneous live cell imaging with motion compensation for uni-axial mechanical straining or compression.
  • Cells can be imaged even during stretch
  • Stretching chamber allows use of commercially available PDMS sheets
  • Cyclic protocols with different characteristics (linear, sinusoidal, sigmoidal) can be programmed
  • Position of the membrane chamber along the stretch axis can be changed
This compact cell strain device makes it possible to perform life cell imaging studies where a rapid image acquisition is required, such as observing changes of the intracellular Ca2+ concentration when cells are mechanically stimulated.