Order Entry
United States
Orders LinkContactUsLinkComponent
Copper etchant

Copper etchant

1 qt.
Catalog # AA44583-R0
Corrosive
Irritant
Supplier #:  044583-R0
Copper etchant
Catalog # AA44583-R0
CAS Number:  
Copper etchant

1 qt.

CAS Number:  
Price
$122.23
Log in for availability
Each (1qt US)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

Physical Form: Liquid Density: 1.19 Appearance: Dark yellow to brown Odor: Acidic UN No.: 3264 MDL No.: MFCD00161912

Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

Synonyms:  Copper etching solution
  • MDL Number:MFCD00161912
  • Density:1.19
  • Storage Temperature:Ambient
  • UN:3264
  • ADR:8,III

Documents

Specifications

Specifications

  • Size:
    1 qt.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.