Pectinase
1 g
Supplier #: J63408-03
Powder
CAS No.: 9032-75-1 Physical Form: Powder Appearance: Off-white to pale brown MDL No.: MFCD00131809
Documents
- Size:1 g
- Cat. No.:AAJ63408-03
- Form:Powder
- Enzyme Name:Pectinase
- CAS No.:9032-75-1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



